missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoro-4-methoxyaniline, 97%

CAS: 363-47-3 | C7H7F2NO | 159.14 g/mol

$340.70 - $1,030.95

Chemical Identifiers

CAS 363-47-3
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD04115910
InChI Key POVSDXPEJZMSEJ-UHFFFAOYSA-N
Synonym 4-amino-2,6-fluoroanisole, benzenamine, 3,5-difluoro-4-methoxy-9ci, 3,5-difluoro-4-methoxy-aniline, benzenamine,3,5-difluoro-4-methoxy, pubchem10104, 3,5-difluoro-4-methxoyaniline, 3,5-difluoro-4-methoxybenzenamine, 3,5-difluoro-4-methoxyphenylamine, 3,5-difluoro-4-methoxybenzeneamine, 3,5-difluoro-4-methoxy-benzenamine
PubChem CID 2783137
IUPAC Name 3,5-difluoro-4-methoxyaniline
SMILES COC1=C(F)C=C(N)C=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAH2677803
View Documents
Thermo Scientific Chemicals
H2677803
1 g N/A
Only null left
 
AAH2677806
View Documents
Thermo Scientific Chemicals
H2677806
5 g N/A
Only null left
 
Description

Description

3,5-Difluoro-4-methoxyaniline is used in conjunction with thermal energy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Difluoro-4-methoxyaniline is used in conjunction with thermal energy.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

363-47-3
159.14
POVSDXPEJZMSEJ-UHFFFAOYSA-N
2783137
COC1=C(F)C=C(N)C=C1F
C7H7F2NO
MFCD04115910
4-amino-2,6-fluoroanisole, benzenamine, 3,5-difluoro-4-methoxy-9ci, 3,5-difluoro-4-methoxy-aniline, benzenamine,3,5-difluoro-4-methoxy, pubchem10104, 3,5-difluoro-4-methxoyaniline, 3,5-difluoro-4-methoxybenzenamine, 3,5-difluoro-4-methoxyphenylamine, 3,5-difluoro-4-methoxybenzeneamine, 3,5-difluoro-4-methoxy-benzenamine
3,5-difluoro-4-methoxyaniline

Specifications

363-47-3
Characteristic
MFCD04115910
UN2811
Insoluble in water.
COC1=C(F)C=C(N)C=C1F
159.14
159.14
3,5-Difluoro-4-methoxyaniline
77°C to 80°C
C7H7F2NO
1 g
4-amino-2,6-fluoroanisole, benzenamine, 3,5-difluoro-4-methoxy-9ci, 3,5-difluoro-4-methoxy-aniline, benzenamine,3,5-difluoro-4-methoxy, pubchem10104, 3,5-difluoro-4-methxoyaniline, 3,5-difluoro-4-methoxybenzenamine, 3,5-difluoro-4-methoxyphenylamine, 3,5-difluoro-4-methoxybenzeneamine, 3,5-difluoro-4-methoxy-benzenamine
POVSDXPEJZMSEJ-UHFFFAOYSA-N
3,5-difluoro-4-methoxyaniline
2783137
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302+H332-H311-H315-H319

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only