Learn More
3,5-Difluoro-4-methoxybenzyl bromide, 97%
CAS: 706786-42-7 | C8H7BrF2O | 237.04 g/mol
$390.14 - $1179.22
Chemical Identifiers
| CAS | 706786-42-7 |
|---|---|
| Molecular Formula | C8H7BrF2O |
| Molecular Weight (g/mol) | 237.04 |
| MDL Number | MFCD04115916 |
| InChI Key | IQTBMXJVMUSGSL-UHFFFAOYSA-N |
| Synonym | 3,5-difluoro-4-methoxybenzyl bromide, 5-bromomethyl-1,3-difluoro-2-methoxybenzene, 4-bromomethyl-2,6-difluoroanisole, benzene, 5-bromomethyl-1,3-difluoro-2-methoxy |
| PubChem CID | 17750664 |
| IUPAC Name | 5-(bromomethyl)-1,3-difluoro-2-methoxybenzene |
| SMILES | COC1=C(F)C=C(CBr)C=C1F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2652603
|
Thermo Scientific Chemicals
H2652603 |
1 g |
Each for $390.14
|
|
|||||
|
AAH2652606
|
Thermo Scientific Chemicals
H2652606 |
5 g |
Each for $1,179.22
|
|
|||||
Description
3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Difluoro-4-methoxybenzyl bromide is useful for research purposes. It is also used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acid and bases.
Chemical Identifiers
| 706786-42-7 | |
| 237.04 | |
| IQTBMXJVMUSGSL-UHFFFAOYSA-N | |
| 17750664 | |
| COC1=C(F)C=C(CBr)C=C1F |
| C8H7BrF2O | |
| MFCD04115916 | |
| 3,5-difluoro-4-methoxybenzyl bromide, 5-bromomethyl-1,3-difluoro-2-methoxybenzene, 4-bromomethyl-2,6-difluoroanisole, benzene, 5-bromomethyl-1,3-difluoro-2-methoxy | |
| 5-(bromomethyl)-1,3-difluoro-2-methoxybenzene |
Specifications
| 706786-42-7 | |
| C8H7BrF2O | |
| 1 g | |
| 3,5-difluoro-4-methoxybenzyl bromide, 5-bromomethyl-1,3-difluoro-2-methoxybenzene, 4-bromomethyl-2,6-difluoroanisole, benzene, 5-bromomethyl-1,3-difluoro-2-methoxy | |
| IQTBMXJVMUSGSL-UHFFFAOYSA-N | |
| 5-(bromomethyl)-1,3-difluoro-2-methoxybenzene | |
| 17750664 | |
| 97% |
| 39°C to 42°C | |
| MFCD04115916 | |
| UN3261 | |
| Insoluble in water. | |
| COC1=C(F)C=C(CBr)C=C1F | |
| 237.04 | |
| 237.04 | |
| 3,5-Difluoro-4-methoxybenzyl bromide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only