missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Difluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

$312.74 - $312.74

Chemical Identifiers

CAS 210532-25-5
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.59
MDL Number MFCD02091378
InChI Key IIQKIICAOXAXEJ-UHFFFAOYSA-N
Synonym 3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03
PubChem CID 2774058
IUPAC Name 3,5-difluorobenzenesulfonyl chloride
SMILES FC1=CC(=CC(F)=C1)S(Cl)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1950706
View Documents
Thermo Scientific Chemicals
L1950706
5 g
Each for $312.74
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

210532-25-5
212.59
IIQKIICAOXAXEJ-UHFFFAOYSA-N
2774058
FC1=CC(=CC(F)=C1)S(Cl)(=O)=O
C6H3ClF2O2S
MFCD02091378
3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03
3,5-difluorobenzenesulfonyl chloride

Specifications

210532-25-5
>110°C (230°F)
MFCD02091378
UN3261
Moisture sensitive
IIQKIICAOXAXEJ-UHFFFAOYSA-N
3,5-difluorobenzenesulfonyl chloride
2774058
97%
57°C to 59°C
C6H3ClF2O2S
5 g
8053561
3,5-difluorobenzenesulphonyl chloride, 3,5-difluorobenzene-1-sulfonyl chloride, 3,5-difluorobenzenesulfonylchloride, 3,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 3,5-difluoro, 3,5-difluoro-benzenesulfonyl chloride, pubchem8463, acmc-209fh1, ksc201g8f, buttpark 27\07-03
FC1=CC(=CC(F)=C1)S(Cl)(=O)=O
212.59
212.6
3,5-Difluorobenzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'