Learn More
3,5-Difluoromandelic acid, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 132741-31-2 |
---|---|
Molecular Formula | C8H6F2O3 |
Molecular Weight (g/mol) | 188.13 |
MDL Number | MFCD00061300 |
InChI Key | PHMLPPFFMSRWBK-UHFFFAOYSA-N |
Synonym | 3,5-difluoromandelic acid, 2-3,5-difluorophenyl-2-hydroxyacetic acid, 3,5-difluorophenyl hydroxy acetic acid, 3,5-difluoro-mandelic acid, 3,5-difluoro mandelic acid, r/s-3,5-difluoromandelic acid, racemic 3,5-difluoro-mandelic acid, 3,5-difluorophenylhydroxyacetic acid, 3,5-difluorophenyl hydroxyacetic acid |
PubChem CID | 588648 |
IUPAC Name | 2-(3,5-difluorophenyl)-2-hydroxyacetic acid |
SMILES | C1=C(C=C(C=C1F)F)C(C(=O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC398480010
|
Thermo Scientific Chemicals
398480010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
132741-31-2 | |
188.13 | |
PHMLPPFFMSRWBK-UHFFFAOYSA-N | |
588648 | |
C1=C(C=C(C=C1F)F)C(C(=O)O)O |
C8H6F2O3 | |
MFCD00061300 | |
3,5-difluoromandelic acid, 2-3,5-difluorophenyl-2-hydroxyacetic acid, 3,5-difluorophenyl hydroxy acetic acid, 3,5-difluoro-mandelic acid, 3,5-difluoro mandelic acid, r/s-3,5-difluoromandelic acid, racemic 3,5-difluoro-mandelic acid, 3,5-difluorophenylhydroxyacetic acid, 3,5-difluorophenyl hydroxyacetic acid | |
2-(3,5-difluorophenyl)-2-hydroxyacetic acid |
Specifications
132741-31-2 | |
100.0 | |
Glass bottle | |
F2C6H3CH(OH)COOH | |
1g | |
PHMLPPFFMSRWBK-UHFFFAOYSA-N | |
2-(3,5-difluorophenyl)-2-hydroxyacetic acid | |
588648 | |
97% |
96.0 | |
96% min. (HPLC) | |
C8H6F2O3 | |
MFCD00061300 | |
3,5-difluoromandelic acid, 2-3,5-difluorophenyl-2-hydroxyacetic acid, 3,5-difluorophenyl hydroxy acetic acid, 3,5-difluoro-mandelic acid, 3,5-difluoro mandelic acid, r/s-3,5-difluoromandelic acid, racemic 3,5-difluoro-mandelic acid, 3,5-difluorophenylhydroxyacetic acid, 3,5-difluorophenyl hydroxyacetic acid | |
C1=C(C=C(C=C1F)F)C(C(=O)O)O | |
188.13 | |
188.13 | |
3, 5-Difluoromandelic acid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only