Learn More
3,5-Dihydroxybenzoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1166436
Description
3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dihydroxybenzoic acid is used in organic synthesis and as intermediate for the pharmaceutical.
Solubility
Soluble in 50 mg/mL, 95% Ethanol.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
3,5-Dihydroxybenzoic acid | |
∼237°C (decomposition) | |
MFCD00002512 | |
2207864 | |
Soluble in 50mg/mL,95% Ethanol. | |
OC(=O)C1=CC(O)=CC(O)=C1 | |
154.12 | |
CHEBI:39912 | |
98% |
99-10-5 | |
C7H6O4 | |
500 g | |
alpha-resorcylic acid, 5-carboxyresorcinol, benzoic acid, 3,5-dihydroxy, unii-2wc5lmo6l1, 3,5-dihydroxy benzoic acid, 3,5-dihydroxy-benzoic acid, 3,5-dhba, 3,5-dihydroxybenzoicacid, a-resorcylic acid, .alpha.-resorcylic acid | |
UYEMGAFJOZZIFP-UHFFFAOYSA-N | |
3,5-dihydroxybenzoic acid | |
7424 | |
154.12 |
Chemical Identifiers
99-10-5 | |
154.12 | |
UYEMGAFJOZZIFP-UHFFFAOYSA-N | |
7424 | |
3,5-dihydroxybenzoic acid |
C7H6O4 | |
MFCD00002512 | |
alpha-resorcylic acid, 5-carboxyresorcinol, benzoic acid, 3,5-dihydroxy, unii-2wc5lmo6l1, 3,5-dihydroxy benzoic acid, 3,5-dihydroxy-benzoic acid, 3,5-dhba, 3,5-dihydroxybenzoicacid, a-resorcylic acid, .alpha.-resorcylic acid | |
CHEBI:39912 | |
OC(=O)C1=CC(O)=CC(O)=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335-H500
EINECSNumber : 202-730-7
RTECSNumber : VH3708000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only