missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethoxybenzaldehyde, 98%, Thermo Scientific Chemicals

$216.31 - $216.31

Chemical Identifiers

CAS 7311-34-4
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00003366
InChI Key VFZRZRDOXPRTSC-UHFFFAOYSA-N
Synonym 3,5-dimethoxy-benzaldehyde, benzaldehyde, 3,5-dimethoxy, 3,5-dimethoxybenzaldehyd, pubchem2645, 3,5 dimethoxybenzaldehyde, acmc-1bhx6, 3,5 dimethoxylbenzaldehyde, 3,5 dimethoxy benzaldehyde, 3,5-dimethoxy benzaldehyde, ghl.pd_mitscher_leg0.694
PubChem CID 81747
IUPAC Name 3,5-dimethoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(OC)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC155220100
SDS View Documents
Thermo Scientific Chemicals
155220100
10 g Glass bottle
Each for $216.31
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7311-34-4
166.18
VFZRZRDOXPRTSC-UHFFFAOYSA-N
81747
COC1=CC(C=O)=CC(OC)=C1
C9H10O3
MFCD00003366
3,5-dimethoxy-benzaldehyde, benzaldehyde, 3,5-dimethoxy, 3,5-dimethoxybenzaldehyd, pubchem2645, 3,5 dimethoxybenzaldehyde, acmc-1bhx6, 3,5 dimethoxylbenzaldehyde, 3,5 dimethoxy benzaldehyde, 3,5-dimethoxy benzaldehyde, ghl.pd_mitscher_leg0.694
3,5-dimethoxybenzaldehyde

Specifications

7311-34-4
100.0
White to Beige
>112°C
98%
C9H10O3
MFCD00003366
08, II, 291
Solubility in water: insoluble
COC1=CC(C=O)=CC(OC)=C1
166.18
166.18
Crystalline Solid
97.5
44°C to 48°C
151°C (16.0 mmHg)
Authentic
Glass bottle
(CH3O)2C6H3CHO
10 g
3,5-dimethoxy-benzaldehyde, benzaldehyde, 3,5-dimethoxy, 3,5-dimethoxybenzaldehyd, pubchem2645, 3,5 dimethoxybenzaldehyde, acmc-1bhx6, 3,5 dimethoxylbenzaldehyde, 3,5 dimethoxy benzaldehyde, 3,5-dimethoxy benzaldehyde, ghl.pd_mitscher_leg0.694
VFZRZRDOXPRTSC-UHFFFAOYSA-N
3,5-dimethoxybenzaldehyde
81747
98%
3, 5-Dimethoxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-772-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only