Learn More
3,5-Dimethoxybenzoic acid, 99%, ACROS Organics™
$51.24 - $51.24
Chemical Identifiers
CAS | 1132-21-4 |
---|---|
Molecular Formula | C9H10O4 |
Molecular Weight (g/mol) | 182.18 |
MDL Number | MFCD00002502 |
InChI Key | IWPZKOJSYQZABD-UHFFFAOYSA-N |
Synonym | benzoic acid, 3,5-dimethoxy, unii-38my78eckk, 3,5-dimethoxy benzoic acid, 38my78eckk, pubchem3121, 3,5-dimethoxybenzoicacid, acmc-1c4zl, 3,5-dimethoxy-benzoic acid, ksc176m0p, benzoicacid, 3,5-dimethoxy |
PubChem CID | 14332 |
IUPAC Name | 3,5-dimethoxybenzoic acid |
SMILES | COC1=CC(=CC(OC)=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC115460050
|
Thermo Scientific Chemicals
115460050 |
5 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1132-21-4 | |
182.18 | |
IWPZKOJSYQZABD-UHFFFAOYSA-N | |
14332 | |
COC1=CC(=CC(OC)=C1)C(O)=O |
C9H10O4 | |
MFCD00002502 | |
benzoic acid, 3,5-dimethoxy, unii-38my78eckk, 3,5-dimethoxy benzoic acid, 38my78eckk, pubchem3121, 3,5-dimethoxybenzoicacid, acmc-1c4zl, 3,5-dimethoxy-benzoic acid, ksc176m0p, benzoicacid, 3,5-dimethoxy | |
3,5-dimethoxybenzoic acid |
Specifications
1132-21-4 | |
100.0 | |
Beige-Pink to White | |
98.5% min. (HPLC) | |
C9H10O4 | |
MFCD00002502 | |
10, 405 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether | |
COC1=CC(=CC(OC)=C1)C(O)=O | |
182.18 | |
182.17 | |
Crystalline Powder |
98.5 | |
182°C to 185°C | |
Authentic | |
Glass bottle | |
(CH3O)2C6H3CO2H | |
5 g | |
benzoic acid, 3,5-dimethoxy, unii-38my78eckk, 3,5-dimethoxy benzoic acid, 38my78eckk, pubchem3121, 3,5-dimethoxybenzoicacid, acmc-1c4zl, 3,5-dimethoxy-benzoic acid, ksc176m0p, benzoicacid, 3,5-dimethoxy | |
IWPZKOJSYQZABD-UHFFFAOYSA-N | |
3,5-dimethoxybenzoic acid | |
14332 | |
98% | |
3, 5-Dimethoxybenzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
May cause respiratory irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective glov
GHS Signal Word: Warning
EINECSNumber : 214-473-8
RUO – Research Use Only