missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethyl-1-adamantaneacetic acid, 97%, Thermo Scientific Chemicals

$150.66 - $529.59

Chemical Identifiers

CAS 14202-14-3
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.328
MDL Number MFCD00077207
InChI Key FUOXJVUIQUYDDI-UHFFFAOYSA-N
Synonym 3,5-dimethyl-1-adamantyl acetic acid, 3,5-dimethyl-1-adamantaneacetic acid, 3,5-dimethyladamantane-1-acetic acid, 2-3,5-dimethyl-1-adamantyl acetic acid, 2-3,5-dimethyladamantan-1-yl acetic acid, 3,5-dimethyladamantan-1-yl acetic acid, 2-3,5-dimethyladamantanyl acetic acid, 3,5-dimethyltricyclo 3.3.1.1∼3,7∼ dec-1-yl acetic acid, acmc-20ancc, 3,5-dimethyladamantaneacetic acid
PubChem CID 4067589
IUPAC Name 2-(3,5-dimethyl-1-adamantyl)acetic acid
SMILES CC12CC3CC(C1)(CC(C3)(C2)CC(=O)O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6635703
View Documents
Thermo Scientific Chemicals
H6635703
1 g
Each for $150.66
 
AAH6635706
View Documents
Thermo Scientific Chemicals
H6635706
5 g
Each for $529.59
 
Description

Description

A commonly used as an organic reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A commonly used as an organic reagent and pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

14202-14-3
222.328
FUOXJVUIQUYDDI-UHFFFAOYSA-N
4067589
CC12CC3CC(C1)(CC(C3)(C2)CC(=O)O)C
C14H22O2
MFCD00077207
3,5-dimethyl-1-adamantyl acetic acid, 3,5-dimethyl-1-adamantaneacetic acid, 3,5-dimethyladamantane-1-acetic acid, 2-3,5-dimethyl-1-adamantyl acetic acid, 2-3,5-dimethyladamantan-1-yl acetic acid, 3,5-dimethyladamantan-1-yl acetic acid, 2-3,5-dimethyladamantanyl acetic acid, 3,5-dimethyltricyclo 3.3.1.1∼3,7∼ dec-1-yl acetic acid, acmc-20ancc, 3,5-dimethyladamantaneacetic acid
2-(3,5-dimethyl-1-adamantyl)acetic acid

Specifications

14202-14-3
White
MFCD00077207
3,5-dimethyl-1-adamantyl acetic acid, 3,5-dimethyl-1-adamantaneacetic acid, 3,5-dimethyladamantane-1-acetic acid, 2-3,5-dimethyl-1-adamantyl acetic acid, 2-3,5-dimethyladamantan-1-yl acetic acid, 3,5-dimethyladamantan-1-yl acetic acid, 2-3,5-dimethyladamantanyl acetic acid, 3,5-dimethyltricyclo 3.3.1.1∼3,7∼ dec-1-yl acetic acid, acmc-20ancc, 3,5-dimethyladamantaneacetic acid
FUOXJVUIQUYDDI-UHFFFAOYSA-N
2-(3,5-dimethyl-1-adamantyl)acetic acid
4067589
97%
3,5-Dimethyl-1-adamantaneacetic acid
110°C to 115°C
C14H22O2
1 g
Slightly soluble in water.
CC12CC3CC(C1)(CC(C3)(C2)CC(=O)O)C
222.328
222.32
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only