Learn More
3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
$169.29 - $566.11
Chemical Identifiers
CAS | 80466-78-0 |
---|---|
Molecular Formula | C5H7ClN2O2S |
Molecular Weight (g/mol) | 194.63 |
MDL Number | MFCD04969910 |
InChI Key | NBGSZCQLPLYKJH-UHFFFAOYSA-N |
Synonym | 1h-pyrazole-4-sulfonylchloride, 3,5-dimethyl, 3,5-dimethylpyrazol-4-yl chlorosulfone, 1h-pyrazole-4-sulfonyl chloride, 3,5-dimethyl, pyrazole, 4-chlorosulfonyl-3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-sulfonylchloride, # |
PubChem CID | 594587 |
IUPAC Name | 3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride |
SMILES | CC1=C(C(C)=NN1)S(Cl)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH5405003
|
Thermo Scientific Chemicals
H5405003 |
1 g |
Each for $169.29
|
|
||||||
AAH5405006
|
Thermo Scientific Chemicals
H5405006 |
5 g |
Each for $566.11
|
|
||||||
Description
3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride is used as pharmaceutical intermediate.
Notes
Moisture sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
80466-78-0 | |
194.63 | |
NBGSZCQLPLYKJH-UHFFFAOYSA-N | |
594587 | |
CC1=C(C(C)=NN1)S(Cl)(=O)=O |
C5H7ClN2O2S | |
MFCD04969910 | |
1h-pyrazole-4-sulfonylchloride, 3,5-dimethyl, 3,5-dimethylpyrazol-4-yl chlorosulfone, 1h-pyrazole-4-sulfonyl chloride, 3,5-dimethyl, pyrazole, 4-chlorosulfonyl-3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-sulfonylchloride, # | |
3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride |
Specifications
80466-78-0 | |
MFCD04969910 | |
UN3261 | |
1h-pyrazole-4-sulfonylchloride, 3,5-dimethyl, 3,5-dimethylpyrazol-4-yl chlorosulfone, 1h-pyrazole-4-sulfonyl chloride, 3,5-dimethyl, pyrazole, 4-chlorosulfonyl-3,5-dimethyl, 3,5-dimethyl-1h-pyrazole-4-sulfonylchloride, # | |
CC1=C(C(C)=NN1)S(Cl)(=O)=O | |
194.63 | |
194.64 | |
3,5-Dimethyl-1H-pyrazole-4-sulfonyl chloride |
C5H7ClN2O2S | |
1 g | |
Moisture sensitive | |
NBGSZCQLPLYKJH-UHFFFAOYSA-N | |
3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride | |
594587 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only