missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dimethyl-4H-1,2,4-triazol-4-amine, 97%, Thermo Scientific™

$294.68 - $294.68

Chemical Identifiers

CAS 3530-15-2
Molecular Formula C4H8N4
Molecular Weight (g/mol) 112.136
MDL Number MFCD00051647
InChI Key MIIKMZAVLKMOFM-UHFFFAOYSA-N
PubChem CID 229800
IUPAC Name 3,5-dimethyl-1,2,4-triazol-4-amine
SMILES CC1=NN=C(N1N)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CD08995EA
SDS View Documents
Thermo Scientific Chemicals
CD08995EA
10 g Amber Glass Bottle
Each for $294.68
 
Description

Description

Specifications

Chemical Identifiers

3530-15-2
112.136
MIIKMZAVLKMOFM-UHFFFAOYSA-N
3,5-dimethyl-1,2,4-triazol-4-amine
C4H8N4
MFCD00051647
229800
CC1=NN=C(N1N)C

Specifications

3530-15-2
100.0
Amber Glass Bottle
MFCD00051647
3,5-dimethyl-4h-1,2,4-triazol-4-amine, 4-amino-3,5-dimethyl-1,2,4-triazole, dimethyl-1,2,4-triazol-4-amine, dimethyl-4h-1,2,4-triazol-4-amine, 4h-1,2,4-triazol-4-amine, 3,5-dimethyl, 3,5-dimethyl-1,2,4-triazole-4-ylamine, acmc-1cpm8, 3,5-dimethyl-4-amino-1,2,4-triazole, 4-amino-3,5-dimethyl-1,2,4 triazole, 4h-1,2,4-triazol-4-amine,3,5-dimethyl
CC1=NN=C(N1N)C
112.136
112.14
3,5-Dimethyl-4H-1,2,4-triazol-4-amine
96.5
97%
C4H8N4
10 g
MIIKMZAVLKMOFM-UHFFFAOYSA-N
3,5-dimethyl-1,2,4-triazol-4-amine
229800
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'