missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylbenzoyl Chloride 97.0+%, TCI America™
Supplier: TCI America D35505G
Specifications
| 3,5-Dimethylbenzoyl Chloride | |
| Yellow | |
| 5 g | |
| C9H9ClO | |
| 3265 | |
| ZJIOBDJEKDUUCI-UHFFFAOYSA-N | |
| 3,5-dimethylbenzoyl chloride | |
| 81088 | |
| ≥97.0% (GC,T) |
| 6613-44-1 | |
| 127°C | |
| Clear Liquid at 20°C | |
| MFCD02313742 | |
| 3,5-dimethyl benzoyl chloride, benzoyl chloride, 3,5-dimethyl, 3,5-dimethylbenzoylchloride, m-xylene-5-carbonyl chloride, pubchem15465, acmc-1b8cp, 5-chloroformyl-m-xylene, 3,5 dimethylbenzoyl chloride, 3.5-dimethylbenzoyl chloride, 5-chlorocarbonyl-m-xylene | |
| CC1=CC(=CC(=C1)C(=O)Cl)C | |
| 168.62 | |
| 168.62 |
Chemical Identifiers
| 6613-44-1 | |
| 168.62 | |
| ZJIOBDJEKDUUCI-UHFFFAOYSA-N | |
| 81088 | |
| CC1=CC(=CC(=C1)C(=O)Cl)C |
| C9H9ClO | |
| MFCD02313742 | |
| 3,5-dimethyl benzoyl chloride, benzoyl chloride, 3,5-dimethyl, 3,5-dimethylbenzoylchloride, m-xylene-5-carbonyl chloride, pubchem15465, acmc-1b8cp, 5-chloroformyl-m-xylene, 3,5 dimethylbenzoyl chloride, 3.5-dimethylbenzoyl chloride, 5-chlorocarbonyl-m-xylene | |
| 3,5-dimethylbenzoyl chloride |
Safety and Handling
EINECSNumber : (3)-4261
RTECSNumber : DM6636900
TSCA : Yes