Learn More
3,5-Dimethylisoxazol-4-yl isocyanate, 95%, Thermo Scientific™
$94.10 - $328.65
Chemical Identifiers
CAS | 131825-41-7 |
---|---|
Molecular Formula | C6H6N2O2 |
Molecular Weight (g/mol) | 138.126 |
MDL Number | MFCD00119910 |
InChI Key | JJQYAGYVYNMYGE-UHFFFAOYSA-N |
Synonym | 3,5-dimethylisoxazol-4-yl isocyanate, isoxazole,4-isocyanato-3,5-dimethyl, 3,5-dimethylisoxazole-4-isocyanate, 4-isocyanato-3,5-dimethylisoxazole, 3,5-dimethyl isoxazol-4-yl isocyanate, 3,5-dimethylisoxazol-4-ylisocyanate, acmc-20mu97, 3,5-dimethylisoxazol-4-isocyanate, 3,5-dimethyl-4-isocyanatoisoxazole, 4-isocyanato3,5-dimethyl-isoxazole |
PubChem CID | 2736266 |
IUPAC Name | 4-isocyanato-3,5-dimethyl-1,2-oxazole |
SMILES | CC1=C(C(=NO1)C)N=C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC00606CB
|
Thermo Scientific Chemicals
CC00606CB |
250 mg | Amber Glass Bottle |
Each for $94.10
|
|
||||
CC00606DA
|
Thermo Scientific Chemicals
CC00606DA |
1 g | Amber Glass Bottle |
Each for $328.65
|
|
||||
Chemical Identifiers
131825-41-7 | |
138.126 | |
JJQYAGYVYNMYGE-UHFFFAOYSA-N | |
2736266 | |
CC1=C(C(=NO1)C)N=C=O |
C6H6N2O2 | |
MFCD00119910 | |
3,5-dimethylisoxazol-4-yl isocyanate, isoxazole,4-isocyanato-3,5-dimethyl, 3,5-dimethylisoxazole-4-isocyanate, 4-isocyanato-3,5-dimethylisoxazole, 3,5-dimethyl isoxazol-4-yl isocyanate, 3,5-dimethylisoxazol-4-ylisocyanate, acmc-20mu97, 3,5-dimethylisoxazol-4-isocyanate, 3,5-dimethyl-4-isocyanatoisoxazole, 4-isocyanato3,5-dimethyl-isoxazole | |
4-isocyanato-3,5-dimethyl-1,2-oxazole |
Specifications
131825-41-7 | |
100.0 | |
Amber Glass Bottle | |
MFCD00119910 | |
3,5-dimethylisoxazol-4-yl isocyanate, isoxazole,4-isocyanato-3,5-dimethyl, 3,5-dimethylisoxazole-4-isocyanate, 4-isocyanato-3,5-dimethylisoxazole, 3,5-dimethyl isoxazol-4-yl isocyanate, 3,5-dimethylisoxazol-4-ylisocyanate, acmc-20mu97, 3,5-dimethylisoxazol-4-isocyanate, 3,5-dimethyl-4-isocyanatoisoxazole, 4-isocyanato3,5-dimethyl-isoxazole | |
CC1=C(C(=NO1)C)N=C=O | |
138.126 | |
138.13 | |
3, 5-Dimethylisoxazol-4-yl isocyanate |
94.5 | |
95% | |
C6H6N2O2 | |
250 mg | |
JJQYAGYVYNMYGE-UHFFFAOYSA-N | |
4-isocyanato-3,5-dimethyl-1,2-oxazole | |
2736266 | |
95% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning