Learn More
3,5-Dimethylphenol, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1223622
Description
3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dimethylphenol finds application as adhesives, sealant chemicals, plating and surface treating agents. It is also used as a solvent for cleaning or degreasing. Further, it is employed as a chemical intermediate for antioxidants, phenolic resins, active pharmaceutical ingredients and metaxalone (muscle relaxant).
Solubility
Soluble in ethyl alcohol, carbon tetrachloride, benzene, chloroform, water and ether.
Notes
Incompatible with bases, acid chlorides, acid anhydrides, oxidizing agents, brass and copper.
Specifications
3,5-Dimethylphenol | |
61°C to 65°C | |
222°C | |
C8H10O | |
100 g | |
774117 | |
3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa | |
TUAMRELNJMMDMT-UHFFFAOYSA-N | |
3,5-dimethylphenol | |
7948 | |
122.17 |
108-68-9 | |
1.12 | |
109°C (228°F) | |
MFCD00002307 | |
UN2261 | |
14,10082 | |
Soluble in ethyl alcohol,carbon tetrachloride,benzene,chloroform,water and ether. | |
CC1=CC(O)=CC(C)=C1 | |
122.17 | |
CHEBI:38572 | |
≥98% |
Chemical Identifiers
108-68-9 | |
122.17 | |
TUAMRELNJMMDMT-UHFFFAOYSA-N | |
7948 | |
3,5-dimethylphenol |
C8H10O | |
MFCD00002307 | |
3,5-xylenol, sym-m-xylenol, phenol, 3,5-dimethyl, 1,3,5-xylenol, 3,5-dmp, 1-hydroxy-3,5-dimethylbenzene, 3,5-dimethyl phenol, 5-hydroxy-m-xylene, xylenol 200, unii-ona760g0wa | |
CHEBI:38572 | |
CC1=CC(O)=CC(C)=C1 |
Safety and Handling
GHS H Statement
H301-H311-H314
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
P260-P264b-P270-P280-P301+P312-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H301+H311-H314
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLENOLS, SOLID
EINECSNumber : 203-606-5
RTECSNumber : ZE6475000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only