missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dinitrobenzamide, 98%, Thermo Scientific Chemicals

$156.85 - $534.93

Chemical Identifiers

CAS 121-81-3
Molecular Formula C7H5N3O5
Molecular Weight (g/mol) 211.13
MDL Number MFCD00007985
InChI Key UUKWKUSGGZNXGA-UHFFFAOYSA-N
Synonym nitromide, tristat, unistat, benzamide, 3,5-dinitro, nitromide usan, component of tristat, nitroamide, component of unistat-3, unii-9duj3cmk8s, 9duj3cmk8s
PubChem CID 4511
IUPAC Name 3,5-dinitrobenzamide
SMILES NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2186514
View Documents
Thermo Scientific Chemicals
B2186514
25 g
Each for $156.85
 
AAB2186522
View Documents
Thermo Scientific Chemicals
B2186522
100 g
Each for $534.93
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-81-3
211.13
UUKWKUSGGZNXGA-UHFFFAOYSA-N
4511
NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
C7H5N3O5
MFCD00007985
nitromide, tristat, unistat, benzamide, 3,5-dinitro, nitromide usan, component of tristat, nitroamide, component of unistat-3, unii-9duj3cmk8s, 9duj3cmk8s
3,5-dinitrobenzamide

Specifications

121-81-3
C7H5N3O5
25 g
nitromide, tristat, unistat, benzamide, 3,5-dinitro, nitromide usan, component of tristat, nitroamide, component of unistat-3, unii-9duj3cmk8s, 9duj3cmk8s
NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
211.13
211.13
3,5-Dinitrobenzamide
181°C to 183°C
MFCD00007985
14,6612
UUKWKUSGGZNXGA-UHFFFAOYSA-N
3,5-dinitrobenzamide
4511
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

EINECSNumber : 204-499-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only