missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1650230

 View more versions of this product

Catalog No. AAA1650230


Explore available promotions

Description

Description

3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dinitrobenzoic acid, is used as a reagent used in the derivatization of resins and the determination of ampicillin.

Solubility
It is Soluble in ethanol (50 mg/mL), methanol, alcohol (very), and glacial acetic acid (very). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases, Strong reducing agents.
Specifications

Specifications

3,5-Dinitrobenzoic acid
204°C to 208°C
C7H4N2O6
250 g
1914286
3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid
VYWYYJYRVSBHJQ-UHFFFAOYSA-N
3,5-dinitrobenzoic acid
7433
212.12
99-34-3
1.68
MFCD00007253
UN1325
14,3276
It is Soluble in ethanol (50mg/mL),methanol,alcohol (very),and glacial acetic acid (very). Insoluble in water.
OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
212.12
CHEBI:73914
≥98%

Chemical Identifiers

99-34-3
212.12
VYWYYJYRVSBHJQ-UHFFFAOYSA-N
7433
3,5-dinitrobenzoic acid
C7H4N2O6
MFCD00007253
3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid
CHEBI:73914
OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H228-H315-H319-H335
Toxic if swallowed.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 202-751-1

RTECSNumber : DG9140700

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only