missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Dinitrobenzoyl Chloride, 99%, Thermo Scientific Chemicals

$77.89 - $207.77

Chemical Identifiers

CAS 99-33-2
Molecular Formula C7H3ClN2O5
Molecular Weight (g/mol) 230.56
MDL Number MFCD00007248
InChI Key NNOHXABAQAGKRZ-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
PubChem CID 7432
IUPAC Name 3,5-dinitrobenzoyl chloride
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC408710250
SDS View Documents
Thermo Scientific Chemicals
408710250
25 g Glass bottle
Each for $77.89
 
AC408711000
SDS View Documents
Thermo Scientific Chemicals
408711000
100 g Glass bottle
Each for $207.77
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

99-33-2
230.56
NNOHXABAQAGKRZ-UHFFFAOYSA-N
7432
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl
C7H3ClN2O5
MFCD00007248
benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
3,5-dinitrobenzoyl chloride

Specifications

99-33-2
100.0
Yellow to Beige
Authentic
Glass bottle
MFCD00007248
09, 414
15, 3303
Solubility in water: decomposes
C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl
230.56
230.56
Crystals or Crystalline Needles
98.5
67.0°C to 70.0°C
196°C (15.0 hPa)
99%
C7H3ClN2O5
25 g
01,320
benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
NNOHXABAQAGKRZ-UHFFFAOYSA-N
3,5-dinitrobenzoyl chloride
7432
99%
3, 5-Dinitrobenzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

EINECSNumber : 202-750-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only