Learn More
3,5-Dinitrosalicylic acid, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1698830
Description
3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Dinitrosalicylic acid is used in the quantification of reducing sugars by colorimetry. It is utilized for the analysis of glycosidase (glycoside hydrolase) activity by quantitation of enzymatically released reducing sugar. It is also used to detect reducing substances in urine and for quantifying carbohydrate level in blood. It acts as an important raw material and an intermediate in the production of organic chemicals such as agrochemicals, organic synthesis, pharmaceuticals and dyes.
Solubility
Soluble in water, dimethyl sulfoxide, methanol, ethanol, diethyl ether and benzene.
Notes
Incompatible with strong oxidizing agents, strong bases, strong reducing agents and strong acids.
Specifications
3,5-Dinitrosalicylic acid | |
169°C to 174°C | |
MFCD00007104 | |
2220661 | |
Soluble in water,dimethyl sulfoxide,methanol,ethanol,diethyl ether and benzene. | |
C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-] | |
11873 | |
228.12 |
609-99-4 | |
C7H4N2O7 | |
250 g | |
3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid | |
LWFUFLREGJMOIZ-UHFFFAOYSA-N | |
228.116 | |
CHEBI:53648 | |
≥97% |
Chemical Identifiers
609-99-4 | |
228.116 | |
LWFUFLREGJMOIZ-UHFFFAOYSA-N | |
11873 | |
C1=C(C=C(C(=C1[N+](=O)[O-])O)C(=O)O)[N+](=O)[O-] |
C7H4N2O7 | |
MFCD00007104 | |
3,5-dinitrosalicylic acid, 2-hydroxy-3,5-dinitrobenzoic acid, 3,5-dinitro-2-hydroxybenzoic acid, 3,5-dinitrosalicylate, benzoic acid, 2-hydroxy-3,5-dinitro, salicylic acid, 3,5-dinitro, dnsa, o-dncp, o-dinitrocarboxylphenol, 3,5-dinitro-2-salicylic acid | |
CHEBI:53648 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 210-204-3
RTECSNumber : VO2965000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only