missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(5-Methyl-2-furyl)butyraldehyde, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2267114

 View more versions of this product

Catalog No. AAB2267114


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(5-Methyl-2-furyl)butyraldehyde
1.006
58°C (136°F)
MFCD00209519
UN1989
3-5-methyl-2-furyl butyraldehyde, 3-5-methylfuran-2-yl butanal, 3-5-methyl-2-furyl butanal, 3-5-methyl-2-furanyl butanal, fema no. 3307, beta5-dimethyl-2-furanpropanal, 2-furanpropanal, beta5-dimethyl, 2-furanpropanal, beta,5-dimethyl, beta5-dimethyl-2-furanpropionaldehyde, 2-furanpropanal, .beta.,5-dimethyl
CCC(C=O)C1=CC=C(C)O1
152.19
152.19
31704-80-0
93°C to 94°C (14 mmHg)
C9H12O2
25 g
Air Sensitive
PFLUFBMKOXFWSD-UHFFFAOYNA-N
3-(5-methylfuran-2-yl)butanal
3578033
99%

Chemical Identifiers

31704-80-0
152.19
PFLUFBMKOXFWSD-UHFFFAOYNA-N
3578033
CCC(C=O)C1=CC=C(C)O1
C9H12O2
MFCD00209519
3-5-methyl-2-furyl butyraldehyde, 3-5-methylfuran-2-yl butanal, 3-5-methyl-2-furyl butanal, 3-5-methyl-2-furanyl butanal, fema no. 3307, beta5-dimethyl-2-furanpropanal, 2-furanpropanal, beta5-dimethyl, 2-furanpropanal, beta,5-dimethyl, beta5-dimethyl-2-furanpropionaldehyde, 2-furanpropanal, .beta.,5-dimethyl
3-(5-methylfuran-2-yl)butanal
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

EINECSNumber : 250-771-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only