missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3(5)-Phenyl-1H-pyrazole, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 296030025

 View more versions of this product

Catalog No. AC296030025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3(5)-Phenyl-1H-pyrazole
99.0
Authentic
Glass bottle
MFCD00159654
23,177
Solubility in water: .. Other solubilities: halogenated solvents,alcohols
N1N=CC=C1C1=CC=CC=C1
144.18
144.18
2458-26-6
100.0
99% min. (GC)
C9H8N2
2.5g
3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
OEDUIFSDODUDRK-UHFFFAOYSA-N
5-phenyl-1H-pyrazole
17155
99+%

Chemical Identifiers

2458-26-6
144.18
OEDUIFSDODUDRK-UHFFFAOYSA-N
17155
N1N=CC=C1C1=CC=CC=C1
C9H8N2
MFCD00159654
3-phenyl-1h-pyrazole, 3-phenylpyrazole, 1h-pyrazole, 3-phenyl, pyrazole, 3-phenyl, 3-benzylpyrazole, 3-phenylpyrazol, phenylpyrazole, 3-phenyl-2h-pyrazole, chembl38876, 5-phenylpyrazole
5-phenyl-1H-pyrazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only