missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[5-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl]propan-1-ol, 97%, Thermo Scientific™

Chemical Identifiers

CAS 175135-15-6
Molecular Formula C11H11F3N2O
Molecular Weight (g/mol) 244.217
MDL Number MFCD00067735
InChI Key FDRMNTVCDKSRNL-UHFFFAOYSA-N
Synonym 3-5-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol, 2-3-hydroxy-1-propyl-5-trifluoromethyl benzimidazole, 3-5-trifluoromethyl-1h-1,3-benzodiazol-2-yl propan-1-ol, 2-3-hydroxy-n-propyl-5-trifluoromethyl-benzimidazole, 3-6-trifluoromethyl-1h-benzimidazol-2-yl propan-1-ol, 2-3-hydroxypropyl-5-trifluoromethyl benzimidazole, 2-3-hydroxy-n-propyl-5-trifluoromethyl benzimidazole, 2-3-hydroxypropyl-5-trifluoromethyl-1h-benzimidazole, 3-6-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol
PubChem CID 2775098
IUPAC Name 3-[6-(trifluoromethyl)-1H-benzimidazol-2-yl]propan-1-ol
SMILES C1=CC2=C(C=C1C(F)(F)F)NC(=N2)CCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB05684DA
SDS View Documents
Thermo Scientific Chemicals
BTB05684DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB05684EA
SDS View Documents
Thermo Scientific Chemicals
BTB05684EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

175135-15-6
244.217
FDRMNTVCDKSRNL-UHFFFAOYSA-N
2775098
C1=CC2=C(C=C1C(F)(F)F)NC(=N2)CCCO
C11H11F3N2O
MFCD00067735
3-5-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol, 2-3-hydroxy-1-propyl-5-trifluoromethyl benzimidazole, 3-5-trifluoromethyl-1h-1,3-benzodiazol-2-yl propan-1-ol, 2-3-hydroxy-n-propyl-5-trifluoromethyl-benzimidazole, 3-6-trifluoromethyl-1h-benzimidazol-2-yl propan-1-ol, 2-3-hydroxypropyl-5-trifluoromethyl benzimidazole, 2-3-hydroxy-n-propyl-5-trifluoromethyl benzimidazole, 2-3-hydroxypropyl-5-trifluoromethyl-1h-benzimidazole, 3-6-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol
3-[6-(trifluoromethyl)-1H-benzimidazol-2-yl]propan-1-ol

Specifications

175135-15-6
100.0
Amber Glass Bottle
MFCD00067735
3-5-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol, 2-3-hydroxy-1-propyl-5-trifluoromethyl benzimidazole, 3-5-trifluoromethyl-1h-1,3-benzodiazol-2-yl propan-1-ol, 2-3-hydroxy-n-propyl-5-trifluoromethyl-benzimidazole, 3-6-trifluoromethyl-1h-benzimidazol-2-yl propan-1-ol, 2-3-hydroxypropyl-5-trifluoromethyl benzimidazole, 2-3-hydroxy-n-propyl-5-trifluoromethyl benzimidazole, 2-3-hydroxypropyl-5-trifluoromethyl-1h-benzimidazole, 3-6-trifluoromethyl-1h-benzo d imidazol-2-yl propan-1-ol
C1=CC2=C(C=C1C(F)(F)F)NC(=N2)CCCO
244.217
244.21
3-[5-(Trifluoromethyl)-1H-benzo[d]imidazol-2-yl]propan-1-ol
96.5
97%
C11H11F3N2O
1g
FDRMNTVCDKSRNL-UHFFFAOYSA-N
3-[6-(trifluoromethyl)-1H-benzimidazol-2-yl]propan-1-ol
2775098
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'