missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[3-(6-Aminohexylamino)propyl]trimethoxysilane 97.0+%, TCI America™
$287.06 - $863.06
Chemical Identifiers
| CAS | 51895-58-0 |
|---|---|
| Molecular Formula | C12H30N2O3Si |
| Molecular Weight (g/mol) | 278.47 |
| MDL Number | MFCD00054943 |
| InChI Key | AMVXVPUHCLLJRE-UHFFFAOYSA-N |
| Synonym | N-[3-(Trimethoxysilyl)propyl]hexamethylenediamine, N1-[3-(Trimethoxysilyl)propyl]hexane-1,6-diamine |
| PubChem CID | 9925638 |
| IUPAC Name | (6-aminohexyl)[3-(trimethoxysilyl)propyl]amine |
| SMILES | CO[Si](CCCNCCCCCCN)(OC)OC |
Chemical Identifiers
| 51895-58-0 | |
| 278.47 | |
| AMVXVPUHCLLJRE-UHFFFAOYSA-N | |
| 9925638 | |
| CO[Si](CCCNCCCCCCN)(OC)OC |
| C12H30N2O3Si | |
| MFCD00054943 | |
| N-[3-(Trimethoxysilyl)propyl]hexamethylenediamine, N1-[3-(Trimethoxysilyl)propyl]hexane-1,6-diamine | |
| (6-aminohexyl)[3-(trimethoxysilyl)propyl]amine |
Specifications
| 51895-58-0 | |
| 165°C | |
| MFCD00054943 | |
| N-[3-(Trimethoxysilyl)propyl]hexamethylenediamine, N1-[3-(Trimethoxysilyl)propyl]hexane-1,6-diamine | |
| CO[Si](CCCNCCCCCCN)(OC)OC | |
| 278.47 | |
| 278.47 | |
| Liquid |
| Yellow | |
| C12H30N2O3Si | |
| 5 mL | |
| AMVXVPUHCLLJRE-UHFFFAOYSA-N | |
| (6-aminohexyl)[3-(trimethoxysilyl)propyl]amine | |
| 9925638 | |
| ≥97.0% (GC,T) | |
| [3-(6-Aminohexylamino)propyl]trimethoxysilane |
Safety and Handling
TSCA : No