missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Bis(hydroxymethyl)durene 99.0+%, TCI America™
$219.95 - $566.67
Chemical Identifiers
| CAS | 7522-62-5 |
|---|---|
| Molecular Formula | C12H18O2 |
| Molecular Weight (g/mol) | 194.27 |
| MDL Number | MFCD00004623 |
| InChI Key | KDJOOHBQJRVMIX-UHFFFAOYSA-N |
| Synonym | 1,4-Bis(hydroxymethyl)-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-p-xylene-alpha,alpha′C-diol |
| PubChem CID | 82026 |
| IUPAC Name | [4-(hydroxymethyl)-2,3,5,6-tetramethylphenyl]methanol |
| SMILES | CC1=C(C)C(CO)=C(C)C(C)=C1CO |
Chemical Identifiers
| 7522-62-5 | |
| 194.27 | |
| KDJOOHBQJRVMIX-UHFFFAOYSA-N | |
| 82026 | |
| CC1=C(C)C(CO)=C(C)C(C)=C1CO |
| C12H18O2 | |
| MFCD00004623 | |
| 1,4-Bis(hydroxymethyl)-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-p-xylene-alpha,alpha′C-diol | |
| [4-(hydroxymethyl)-2,3,5,6-tetramethylphenyl]methanol |
Specifications
| 7522-62-5 | |
| C12H18O2 | |
| 1 g | |
| KDJOOHBQJRVMIX-UHFFFAOYSA-N | |
| [4-(hydroxymethyl)-2,3,5,6-tetramethylphenyl]methanol | |
| 82026 | |
| ≥99.0% (GC) | |
| 3,6-Bis(hydroxymethyl)durene |
| 256°C | |
| MFCD00004623 | |
| 1,4-Bis(hydroxymethyl)-2,3,5,6-tetramethylbenzene, 2,3,5,6-Tetramethyl-p-xylene-alpha,alpha′C-diol | |
| CC1=C(C)C(CO)=C(C)C(C)=C1CO | |
| 194.27 | |
| 194.27 | |
| Crystalline Powder |
Safety and Handling
TSCA : No