missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-2-chloropyridine, 95%, Thermo Scientific Chemicals

$293.04 - $293.04

Chemical Identifiers

CAS 942206-18-0
Molecular Formula C5H2Br2ClN
Molecular Weight (g/mol) 271.336
MDL Number MFCD09037464
InChI Key CVZSNXGMWIGPBF-UHFFFAOYSA-N
Synonym 2-chloro-3,6-dibromopyridine, 3,6-dibromo-2-chloro-pyridine
PubChem CID 42553063
IUPAC Name 3,6-dibromo-2-chloropyridine
SMILES C1=CC(=NC(=C1Br)Cl)Br
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2773803
View Documents
Thermo Scientific Chemicals
H2773803
1 g
Each for $293.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

942206-18-0
271.336
CVZSNXGMWIGPBF-UHFFFAOYSA-N
42553063
C1=CC(=NC(=C1Br)Cl)Br
C5H2Br2ClN
MFCD09037464
2-chloro-3,6-dibromopyridine, 3,6-dibromo-2-chloro-pyridine
3,6-dibromo-2-chloropyridine

Specifications

942206-18-0
C5H2Br2ClN
1 g
CVZSNXGMWIGPBF-UHFFFAOYSA-N
3,6-dibromo-2-chloropyridine
42553063
95%
84°C to 86°C
MFCD09037464
2-chloro-3,6-dibromopyridine, 3,6-dibromo-2-chloro-pyridine
C1=CC(=NC(=C1Br)Cl)Br
271.336
271.34
3,6-Dibromo-2-chloropyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only