missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole 95.0+%, TCI America™
$150.98 - $454.83
Chemical Identifiers
| CAS | 946491-48-1 |
|---|---|
| Molecular Formula | C26H27Br2NO |
| Molecular Weight (g/mol) | 529.32 |
| MDL Number | MFCD30721573 |
| InChI Key | ACUVALSRGHGNLZ-UHFFFAOYNA-N |
| PubChem CID | 68106047 |
| IUPAC Name | 3,6-dibromo-9-{4-[(2-ethylhexyl)oxy]phenyl}-9H-carbazole |
| SMILES | CCCCC(CC)COC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D5342200MG
|
TCI America
D5342200MG |
200 mg |
Each for $150.98
|
|
|||||
|
D53421G
|
TCI America
D53421G |
1 g |
Each for $454.83
|
|
|||||
Chemical Identifiers
| 946491-48-1 | |
| 529.32 | |
| ACUVALSRGHGNLZ-UHFFFAOYNA-N | |
| 3,6-dibromo-9-{4-[(2-ethylhexyl)oxy]phenyl}-9H-carbazole |
| C26H27Br2NO | |
| MFCD30721573 | |
| 68106047 | |
| CCCCC(CC)COC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2 |
Specifications
| 946491-48-1 | |
| C26H27Br2NO | |
| 200 mg | |
| CCCCC(CC)COC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2 | |
| 529.32 | |
| 529.32 | |
| Crystalline Powder |
| 59°C | |
| MFCD30721573 | |
| ACUVALSRGHGNLZ-UHFFFAOYNA-N | |
| 3,6-dibromo-9-{4-[(2-ethylhexyl)oxy]phenyl}-9H-carbazole | |
| 68106047 | |
| ≥95.0% (HPLC,N) | |
| 3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole |
Safety and Handling
TSCA : No