missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,6-Dihydroxyflavone, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2698906

 View more versions of this product

Catalog No. AAH2698906


Explore available promotions

Description

Description

3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications

Specifications

3,6-Dihydroxyflavone
231°C to 234°C
MFCD00143082
3,6-dihydroxyflavone, 3,6-dihydroxy-2-phenyl-4h-chromen-4-one, 6-hydroxyflavonol, 3,6-dihydroxy-flavone, 4h-1-benzopyran-4-one, 3,6-dihydroxy-2-phenyl, acmc-1c7fz, 3,6-dihydroxy-2-phenylchromone
XHLOLFKZCUCROE-UHFFFAOYSA-N
3,6-dihydroxy-2-phenylchromen-4-one
688659
97%
108238-41-1
C15H10O4
5 g
Soluble in water.
C1=CC=C(C=C1)C2=C(C(=O)C3=C(O2)C=CC(=C3)O)O
254.241
254.24

Chemical Identifiers

108238-41-1
254.241
XHLOLFKZCUCROE-UHFFFAOYSA-N
688659
C1=CC=C(C=C1)C2=C(C(=O)C3=C(O2)C=CC(=C3)O)O
C15H10O4
MFCD00143082
3,6-dihydroxyflavone, 3,6-dihydroxy-2-phenyl-4h-chromen-4-one, 6-hydroxyflavonol, 3,6-dihydroxy-flavone, 4h-1-benzopyran-4-one, 3,6-dihydroxy-2-phenyl, acmc-1c7fz, 3,6-dihydroxy-2-phenylchromone
3,6-dihydroxy-2-phenylchromen-4-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only