missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(9-Anthryl)propionic acid, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3418403

 View more versions of this product

Catalog No. AAH3418403


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(9-Anthryl)propionic acid
193°C to 197°C
MFCD00068641
9-anthracenepropionic acid, 3-anthracen-9-yl propanoic acid, 3-anthracen-9-yl-propionic acid, 9-anthracenepropanoic acid, 3-9-anthryl propanoic acid, 9apa, 9-anthracenepropanoicacid, 9-an-thracenepropionic acid, 3-9-anthryl-propionic acid, 3-anthracen-9-ylpropionic acid
OC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12
250.30
250.3
41034-83-7
C17H14O2
1 g
BDGMYCZUEIGHJH-UHFFFAOYSA-N
3-anthracen-9-ylpropanoic acid
170457
96%

Chemical Identifiers

41034-83-7
250.30
BDGMYCZUEIGHJH-UHFFFAOYSA-N
170457
OC(=O)CCC1=C2C=CC=CC2=CC2=CC=CC=C12
C17H14O2
MFCD00068641
9-anthracenepropionic acid, 3-anthracen-9-yl propanoic acid, 3-anthracen-9-yl-propionic acid, 9-anthracenepropanoic acid, 3-9-anthryl propanoic acid, 9apa, 9-anthracenepropanoicacid, 9-an-thracenepropionic acid, 3-9-anthryl-propionic acid, 3-anthracen-9-ylpropionic acid
3-anthracen-9-ylpropanoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only