missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane 95.0+%, TCI America™

Supplier:  TCI America B566425G

 View more versions of this product

Catalog No. B566425G

Explore available promotions

Description

Description

Specifications

Specifications

3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
177°C
C33H50O6P2
25 g
AIBRSVLEQRWAEG-FJGSWBKINA-N
3,9-bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
93101
≥95.0% (GC)
26741-53-7
White-Yellow
MFCD00071521
Pentaerythritol Bis(2,4-di-tert-butylphenylphosphite)
CC(C)(C)C1=CC=C(O[P@]2OC[C@]3(CO[P@](OC4=CC=C(C=C4C(C)(C)C)C(C)(C)C)OC3)CO2)C(=C1)C(C)(C)C
604.71
604.70
Crystalline Powder

Chemical Identifiers

26741-53-7
604.71
AIBRSVLEQRWAEG-FJGSWBKINA-N
93101
CC(C)(C)C1=CC=C(O[P@]2OC[C@]3(CO[P@](OC4=CC=C(C=C4C(C)(C)C)C(C)(C)C)OC3)CO2)C(=C1)C(C)(C)C
C33H50O6P2
MFCD00071521
Pentaerythritol Bis(2,4-di-tert-butylphenylphosphite)
3,9-bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-5768

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions