missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Acetamidobenzoic acid, 98%, Thermo Scientific Chemicals

$53.33 - $150.84

Chemical Identifiers

CAS 587-48-4
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00013983
InChI Key RGDPZMQZWZMONQ-UHFFFAOYSA-N
Synonym 3-acetylamino benzoic acid, benzoic acid, 3-acetylamino, 3-acetylaminobenzoic acid, m-acetylaminobenzoic acid, benzoic acid, m-acetamido, n-acetyl-m-aminobenzoic acid, 3-acetylamino-benzoic acid, 3-acetylamino-benzoicaci, 3-acetamidobenzoicacid, 3'-carboxyacetanilide
PubChem CID 48847
IUPAC Name 3-acetamidobenzoic acid
SMILES CC(=O)NC1=CC=CC(=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1590214
View Documents
Thermo Scientific Chemicals
A1590214
25 g
Each for $53.33
 
AAA1590222
View Documents
Thermo Scientific Chemicals
A1590222
100 g
Each for $150.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

587-48-4
179.175
RGDPZMQZWZMONQ-UHFFFAOYSA-N
48847
CC(=O)NC1=CC=CC(=C1)C(=O)O
C9H9NO3
MFCD00013983
3-acetylamino benzoic acid, benzoic acid, 3-acetylamino, 3-acetylaminobenzoic acid, m-acetylaminobenzoic acid, benzoic acid, m-acetamido, n-acetyl-m-aminobenzoic acid, 3-acetylamino-benzoic acid, 3-acetylamino-benzoicaci, 3-acetamidobenzoicacid, 3'-carboxyacetanilide
3-acetamidobenzoic acid

Specifications

587-48-4
C9H9NO3
25 g
3-acetylamino benzoic acid, benzoic acid, 3-acetylamino, 3-acetylaminobenzoic acid, m-acetylaminobenzoic acid, benzoic acid, m-acetamido, n-acetyl-m-aminobenzoic acid, 3-acetylamino-benzoic acid, 3-acetylamino-benzoicaci, 3-acetamidobenzoicacid, 3'-carboxyacetanilide
CC(=O)NC1=CC=CC(=C1)C(=O)O
179.175
179.18
3-Acetamidobenzoic acid
249°C to 252°C
MFCD00013983
2210867
RGDPZMQZWZMONQ-UHFFFAOYSA-N
3-acetamidobenzoic acid
48847
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 209-600-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only