missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Acetylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$131.49 - $131.49
Specifications
CAS | 204841-19-0 |
---|---|
Melting Point | 208°C |
Color | Yellow |
Molecular Formula | C8H9BO3 |
MDL Number | MFCD01074678 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
A19425G
|
TCI America
A19425G |
5 g |
Each for $131.49
|
|
|||||
Specifications
204841-19-0 | |
Yellow | |
MFCD01074678 | |
B(C1=CC(=CC=C1)C(=O)C)(O)O | |
163.967 | |
CHEBI:86552 | |
Crystalline Powder |
208°C | |
C8H9BO3 | |
SJGGDZCTGBKBCK-UHFFFAOYSA-N | |
(3-acetylphenyl)boronic acid | |
2734310 | |
163.97 |