Learn More
3-Amino-1-propanol, 99%, Thermo Scientific Chemicals
$40.66 - $116.52
Chemical Identifiers
CAS | 156-87-6 |
---|---|
MDL Number | MFCD00008223 |
InChI Key | WUGQZFFCHPXWKQ-UHFFFAOYSA-N |
Synonym | 3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol |
PubChem CID | 9086 |
IUPAC Name | 3-aminopropan-1-ol |
SMILES | C(CN)CO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC104460050
|
Thermo Scientific Chemicals
104460050 |
5 mL | Glass bottle |
Each for $40.66
|
|
||||
AC104465000
|
Thermo Scientific Chemicals
104465000 |
500 mL | Glass bottle |
Each for $116.52
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
156-87-6 | |
WUGQZFFCHPXWKQ-UHFFFAOYSA-N | |
9086 | |
C(CN)CO |
MFCD00008223 | |
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol | |
3-aminopropan-1-ol |
Specifications
156-87-6 | |
100.0 | |
Colorless to Yellow | |
187.0°C to 188.0°C | |
Authentic | |
Glass bottle | |
1.4595 to 1.4615 | |
MFCD00008223 | |
04, 288 | |
3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol | |
WUGQZFFCHPXWKQ-UHFFFAOYSA-N | |
3-aminopropan-1-ol | |
9086 | |
75.11 | |
Liquid |
98.5 | |
10.0°C to 12.0°C | |
0.9820g/mL | |
101°C | |
98.5% min. (GC) | |
C3H9NO | |
H2N(CH2)3OH | |
5 mL | |
0.982 | |
Solubility in water: freely soluble. Other solubilities: miscible with alcohols,esters, chloroform, acetone, slightly soluble in arom. and aliph. hydrocarbons | |
C(CN)CO | |
75.11 | |
36 mPa.s (20°C) | |
99% | |
3-Amino-1-propanol, 99% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 205-864-4
RTECSNumber : UA5600000
TSCA : TSCA
RUO – Research Use Only