Learn More
3-Amino-1H-1,2,4-triazole-5-carboxylic acid hydrate, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 1266615-63-7 |
---|---|
Molecular Formula | C3H4N4O2·xH2O |
Molecular Weight (g/mol) | 128.09 |
MDL Number | MFCD00149595 |
Synonym | 5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC249301000
|
Thermo Scientific Chemicals
249301000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1266615-63-7 | |
128.09 | |
5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate |
C3H4N4O2·xH2O | |
MFCD00149595 |
Specifications
1266615-63-7,3641-13-2 | |
Authentic | |
Glass bottle | |
MFCD00149595 | |
5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid hydrate | |
128.09 | |
98% |
2% max. | |
98% | |
C3H4N4O2·xH2O | |
100g | |
Solubility in water: .. Other solubilities: insoluble in dil. acid- soluble in aq. alkalis | |
128.09 | |
3-Amino-1H-1, 2, 4-triazole-5-carboxylic acid hydrate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only