missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-1H-1,2,4-triazole, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1487130

 View more versions of this product

Catalog No. AAA1487130


Explore available promotions

Description

Description

3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1,2,4-triazole is an inhibitor of mitochondrial and chloroplast function. Commercial grade 3- amino-1,2,4-triazole (which generally contains catalase anti-inhibitory impurities) is used as a cotton defoliant.

Solubility
It is soluble in water, methanol, ethanol and chloroform, and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.

Notes
Store desiccated at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from Strong oxidizing agents, Strong acids, Copper, Iron and iron salts.
Specifications

Specifications

3-Amino-1, 2, 4-triazole
147°C to 153°C
96%
MFCD00005230,MFCD00053362
UN3077
14,489
It is soluble in water,methanol,ethanol and chloroform,and sparingly soluble in ethyl acetate. It is insoluble in ether and acetone.
NC1=NC=NN1
84.08
CHEBI:40036
96%
61-82-5
1.138
C2H4N4
250 g
107687
3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
KLSJWNVTNUYHDU-UHFFFAOYSA-N
1H-1,2,4-triazol-5-amine
1639
84.08

Chemical Identifiers

61-82-5
84.08
KLSJWNVTNUYHDU-UHFFFAOYSA-N
1639
1H-1,2,4-triazol-5-amine
C2H4N4
MFCD00005230,MFCD00053362
3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox
CHEBI:40036
NC1=NC=NN1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H373

P201-P202-P260-P264b-P270-P280i-P281-P301+P312-P305+P351+P338-P308+P313-P330-P501c

H302-H319-H350-H360-H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 200-521-5

RTECSNumber : XZ3850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only