missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-2,2-dimethyl-1-propanol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 457430050

 View more versions of this product

Catalog No. AC457430050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

3-Amino-2,2-dimethyl-1-propanol
Authentic
C5H13NO
3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
CC(C)(CN)CO
103.17
103.17
26734-09-8
96% min. (GC)
5g
FNVOFDGAASRDQY-UHFFFAOYSA-N
3-amino-2,2-dimethylpropan-1-ol
117326
97%

Chemical Identifiers

26734-09-8
103.17
3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
3-amino-2,2-dimethylpropan-1-ol
C5H13NO
FNVOFDGAASRDQY-UHFFFAOYSA-N
117326
CC(C)(CN)CO
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 247-948-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only