missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-2-cyclohexen-1-one 98.0+%, TCI America™
$88.87 - $264.65
Chemical Identifiers
CAS | 5220-49-5 |
---|---|
Molecular Formula | C6H9NO |
Molecular Weight (g/mol) | 111.14 |
MDL Number | MFCD00013783 |
InChI Key | ZZMRPOAHZITKBV-UHFFFAOYSA-N |
Synonym | 3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd |
PubChem CID | 78892 |
IUPAC Name | 3-aminocyclohex-2-en-1-one |
SMILES | NC1=CC(=O)CCC1 |
Chemical Identifiers
5220-49-5 | |
111.14 | |
ZZMRPOAHZITKBV-UHFFFAOYSA-N | |
78892 | |
NC1=CC(=O)CCC1 |
C6H9NO | |
MFCD00013783 | |
3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd | |
3-aminocyclohex-2-en-1-one |
Specifications
5220-49-5 | |
C6H9NO | |
5 g | |
ZZMRPOAHZITKBV-UHFFFAOYSA-N | |
3-aminocyclohex-2-en-1-one | |
78892 | |
≥98.0% (GC) | |
3-Amino-2-cyclohexen-1-one |
132°C | |
MFCD00013783 | |
3-amino-2-cyclohexen-1-one, 3-aminocyclohex-2-enone, 2-cyclohexen-1-one, 3-amino, unii-v596547zke, 3-amino-2-cyclohexen-1-ketone, 3-amino-2-cyclohexenone, 3-aminocyclohexenone, 3-aminocyclohex-enone, pubchem14449, acmc-209kyd | |
NC1=CC(=O)CCC1 | |
111.14 | |
111.14 | |
Crystalline Powder |
Safety and Handling
TSCA : No