missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-2-methylbenzoic Acid 99.0+%, TCI America™
$80.97 - $477.95
Chemical Identifiers
CAS | 52130-17-3 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.165 |
MDL Number | MFCD00075026 |
InChI Key | BYHMLZGICSEKIY-UHFFFAOYSA-N |
Synonym | 2-methyl-3-aminobenzoic acid, 3-amino-o-toluic acid, 3-amino-2-methylbenzoicacid, 3-amino-2-methyl-benzoic acid, benzoic acid, 3-amino-2-methyl, 2-methyl-3-amino benzoic acid, pubchem10921, acmc-209ky1, ksc489o4b, 3-amino-2-methyl benzoic acid |
PubChem CID | 2733699 |
IUPAC Name | 3-amino-2-methylbenzoic acid |
SMILES | CC1=C(C=CC=C1N)C(=O)O |
Chemical Identifiers
52130-17-3 | |
151.165 | |
BYHMLZGICSEKIY-UHFFFAOYSA-N | |
2733699 | |
CC1=C(C=CC=C1N)C(=O)O |
C8H9NO2 | |
MFCD00075026 | |
2-methyl-3-aminobenzoic acid, 3-amino-o-toluic acid, 3-amino-2-methylbenzoicacid, 3-amino-2-methyl-benzoic acid, benzoic acid, 3-amino-2-methyl, 2-methyl-3-amino benzoic acid, pubchem10921, acmc-209ky1, ksc489o4b, 3-amino-2-methyl benzoic acid | |
3-amino-2-methylbenzoic acid |
Specifications
52130-17-3 | |
C8H9NO2 | |
25 g | |
BYHMLZGICSEKIY-UHFFFAOYSA-N | |
3-amino-2-methylbenzoic acid | |
2733699 | |
≥99.0% (HPLC,T) | |
3-Amino-2-methylbenzoic Acid |
188°C | |
MFCD00075026 | |
2-methyl-3-aminobenzoic acid, 3-amino-o-toluic acid, 3-amino-2-methylbenzoicacid, 3-amino-2-methyl-benzoic acid, benzoic acid, 3-amino-2-methyl, 2-methyl-3-amino benzoic acid, pubchem10921, acmc-209ky1, ksc489o4b, 3-amino-2-methyl benzoic acid | |
CC1=C(C=CC=C1N)C(=O)O | |
151.165 | |
151.17 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-4438
TSCA : No