missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-3-methyloxetane, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H57588MD

 View more versions of this product

Catalog No. AAH57588MD


Explore available promotions

Description

Description

It is used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-Amino-3-methyloxetane
0.99 g/mL
C4H9NO
250 mg
Slightly soluble in water.
CC1(COC1)N
87.122
87.12
Crystalline Solid
874473-14-0
102°C
MFCD11111713
3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane
NQVWMPOQWBDSAI-UHFFFAOYSA-N
3-methyloxetan-3-amine
46835725
95%

Chemical Identifiers

874473-14-0
87.122
NQVWMPOQWBDSAI-UHFFFAOYSA-N
46835725
CC1(COC1)N
C4H9NO
MFCD11111713
3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane
3-methyloxetan-3-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only