missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Amino-3-methyloxetane, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H57588ME
Description
It is used as an OLED intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an OLED intermediate.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
3-Amino-3-methyloxetane | |
0.99 g/mL | |
C4H9NO | |
500 mg | |
Slightly soluble in water. | |
CC1(COC1)N | |
87.122 | |
87.12 | |
Crystalline Solid |
874473-14-0 | |
102°C | |
MFCD11111713 | |
3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane | |
NQVWMPOQWBDSAI-UHFFFAOYSA-N | |
3-methyloxetan-3-amine | |
46835725 | |
95% |
Chemical Identifiers
874473-14-0 | |
87.122 | |
NQVWMPOQWBDSAI-UHFFFAOYSA-N | |
46835725 | |
CC1(COC1)N |
C4H9NO | |
MFCD11111713 | |
3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane | |
3-methyloxetan-3-amine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only