missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-3-phenyl-1-propanol, 94%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 292935000

 View more versions of this product

Catalog No. AC292935000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Amino-3-phenyl-1-propanol
Authentic
Glass bottle
C6H5CH(NH2)CH2CH2OH
500mg
3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0
SEQXIQNPMQTBGN-UHFFFAOYNA-N
3-amino-3-phenylpropan-1-ol
203500
94%
14593-04-5
94%
C9H13NO
MFCD00801124,MFCD01311768,MFCD01311791
13,IV,1919
Solubility in water: .. Other solubilities: soluble in methanol, ethanol and chloroform
NC(CCO)C1=CC=CC=C1
151.21
151.21

Chemical Identifiers

14593-04-5
151.21
SEQXIQNPMQTBGN-UHFFFAOYNA-N
203500
NC(CCO)C1=CC=CC=C1
C9H13NO
MFCD00801124,MFCD01311768,MFCD01311791
3-amino-3-phenyl-1-propanol, dl-beta-phenylalaninol, 3-amino-3-phenyl-propan-1-ol, 3-phenyl-dl-beta-alaninol, 1-propanol, 3-amino-3-phenyl, benzenepropanol, gamma-amino, 1-amino-3-hydroxyprop-1-yl benzene, dl--phenylalaninol, 3-amino-3-phenylpropanol, acmc-1c4c0
3-amino-3-phenylpropan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only