Learn More
3-Amino-4-methoxybenzoic acid, 98+%, Thermo Scientific Chemicals
$70.32 - $266.33
Chemical Identifiers
CAS | 2840-26-8 |
---|---|
Molecular Formula | C8H9NO3 |
Molecular Weight (g/mol) | 167.164 |
MDL Number | MFCD00002521 |
InChI Key | FDGAEAYZQQCBRN-UHFFFAOYSA-N |
Synonym | 3-amino-p-anisic acid, p-anisic acid, 3-amino, 3-amino-4-methoxy-benzoic acid, 3-amino-4-anisic acid, 3-amino-4-methoxybenzoicacid, benzoic acid, 3-amino-4-methoxy, kyselina 3-amino-4-methoxybenzoova, 3-amino-4-methoxybenzoic, 5-carboxy-2-methoxyaniline, kyselina 3-amino-4-methoxybenzoova czech |
PubChem CID | 17823 |
IUPAC Name | 3-amino-4-methoxybenzoic acid |
SMILES | COC1=C(C=C(C=C1)C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2066906
|
Thermo Scientific Chemicals
B2066906 |
5 g |
Each for $70.32
|
|
|||||
AAB2066914
|
Thermo Scientific Chemicals
B2066914 |
25 g |
Each for $266.33
|
|
|||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
2840-26-8 | |
167.164 | |
FDGAEAYZQQCBRN-UHFFFAOYSA-N | |
17823 | |
COC1=C(C=C(C=C1)C(=O)O)N |
C8H9NO3 | |
MFCD00002521 | |
3-amino-p-anisic acid, p-anisic acid, 3-amino, 3-amino-4-methoxy-benzoic acid, 3-amino-4-anisic acid, 3-amino-4-methoxybenzoicacid, benzoic acid, 3-amino-4-methoxy, kyselina 3-amino-4-methoxybenzoova, 3-amino-4-methoxybenzoic, 5-carboxy-2-methoxyaniline, kyselina 3-amino-4-methoxybenzoova czech | |
3-amino-4-methoxybenzoic acid |
Specifications
2840-26-8 | |
C8H9NO3 | |
5 g | |
Soluble in water. | |
COC1=C(C=C(C=C1)C(=O)O)N | |
167.164 | |
167.16 | |
3-Amino-4-methoxybenzoic acid |
208°C to 210°C | |
MFCD00002521 | |
3-amino-p-anisic acid, p-anisic acid, 3-amino, 3-amino-4-methoxy-benzoic acid, 3-amino-4-anisic acid, 3-amino-4-methoxybenzoicacid, benzoic acid, 3-amino-4-methoxy, kyselina 3-amino-4-methoxybenzoova, 3-amino-4-methoxybenzoic, 5-carboxy-2-methoxyaniline, kyselina 3-amino-4-methoxybenzoova czech | |
FDGAEAYZQQCBRN-UHFFFAOYSA-N | |
3-amino-4-methoxybenzoic acid | |
17823 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-634-3
RTECSNumber : DG2872000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only