missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-4-methylbiphenyl, 98%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals H2826503

Catalog No. AAH2826503

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Amino-4-methylbiphenyl
C13H13N
1 g
5-phenyl-o-toluidine, 3-amino-4-methylbiphenyl, 4-methyl-1,1'-biphenyl-3-amine, 2-methyl-5-phenylphenylamine, 1,1'-biphenyl-3-amine, 4-methyl, 4-methylbiphenyl-3-amine, 2-methyl-5-phenyl-aniline
CC1=CC=C(C=C1N)C1=CC=CC=C1
183.25
183.26
80938-67-6
MFCD00270126
Air Sensitive
YLKSTPDTTKOSIL-UHFFFAOYSA-N
2-methyl-5-phenylaniline
149880
98%

Chemical Identifiers

80938-67-6
183.25
YLKSTPDTTKOSIL-UHFFFAOYSA-N
149880
CC1=CC=C(C=C1N)C1=CC=CC=C1
C13H13N
MFCD00270126
5-phenyl-o-toluidine, 3-amino-4-methylbiphenyl, 4-methyl-1,1'-biphenyl-3-amine, 2-methyl-5-phenylphenylamine, 1,1'-biphenyl-3-amine, 4-methyl, 4-methylbiphenyl-3-amine, 2-methyl-5-phenyl-aniline
2-methyl-5-phenylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

P201-P202-P281-P308+P313-P501c

H351

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only