missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-6-bromo-2-chloropyridine 98.0+%, TCI America™
$61.51 - $180.81
Chemical Identifiers
| CAS | 169833-70-9 |
|---|---|
| Molecular Formula | C5H4BrClN2 |
| Molecular Weight (g/mol) | 207.455 |
| MDL Number | MFCD04971930 |
| InChI Key | YCECOMBEVDPUJG-UHFFFAOYSA-N |
| PubChem CID | 2050085 |
| IUPAC Name | 6-bromo-2-chloropyridin-3-amine |
| SMILES | C1=CC(=NC(=C1N)Cl)Br |
Chemical Identifiers
| 169833-70-9 | |
| 207.455 | |
| YCECOMBEVDPUJG-UHFFFAOYSA-N | |
| 6-bromo-2-chloropyridin-3-amine |
| C5H4BrClN2 | |
| MFCD04971930 | |
| 2050085 | |
| C1=CC(=NC(=C1N)Cl)Br |
Specifications
| 169833-70-9 | |
| C5H4BrClN2 | |
| 1 g | |
| C1=CC(=NC(=C1N)Cl)Br | |
| 207.455 | |
| 207.46 | |
| Crystalline Powder |
| 119°C | |
| MFCD04971930 | |
| YCECOMBEVDPUJG-UHFFFAOYSA-N | |
| 6-bromo-2-chloropyridin-3-amine | |
| 2050085 | |
| ≥98.0% (GC) | |
| 3-Amino-6-bromo-2-chloropyridine |
Safety and Handling
TSCA : No