missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Amino-6-chloropyridazine, 98%, Thermo Scientific Chemicals

$69.40 - $1040.92

Chemical Identifiers

CAS 5469-69-2
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
MDL Number MFCD00051506
InChI Key DTXVKPOKPFWSFF-UHFFFAOYSA-N
Synonym 3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
PubChem CID 21643
IUPAC Name 6-chloropyridazin-3-amine
SMILES NC1=CC=C(Cl)N=N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1018503
View Documents
Thermo Scientific Chemicals
A1018503
1 g
Each for $69.40
 
AAA1018506
View Documents
Thermo Scientific Chemicals
A1018506
5 g
Each for $246.53
 
AAA1018514
View Documents
Thermo Scientific Chemicals
A1018514
25 g
Each for $1,040.92
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5469-69-2
129.55
DTXVKPOKPFWSFF-UHFFFAOYSA-N
21643
NC1=CC=C(Cl)N=N1
C4H4ClN3
MFCD00051506
3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
6-chloropyridazin-3-amine

Specifications

5469-69-2
C4H4ClN3
1 g
3-amino-6-chloropyridazine, 3-pyridazinamine, 6-chloro, 6-chloro-3-pyridazinamine, 6-amino-3-chloropyridazine, 6-chloro-3-aminopyridazine, 6-chloropyridazin-3-ylamine, 6-chloro-pyridazin-3-ylamine, unii-ijs443slzg, 3-chloro-6-pyridazinamine, 6-chloro-3-pyridazinylamine
NC1=CC=C(Cl)N=N1
129.55
129.55
3-Amino-6-chloropyridazine
∼218°C (decomposition)
MFCD00051506
110918
DTXVKPOKPFWSFF-UHFFFAOYSA-N
6-chloropyridazin-3-amine
21643
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 226-796-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only