missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

3-Aminoazetidine dihydrochloride, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H59318ME

 Afficher plus de versions de ce produit

Numéro de catalogue. AAH59318ME


Découvrez plus d'offres spéciales

Description

Description

3-Aminoazetidine dihydrochloride, is used as active pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminoazetidine dihydrochloride, is used as active pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Spécifications

Spécifications

3-Aminoazetidine dihydrochloride
C3H8N2
500 mg
3-aminoazetidine, 3-azetidinamine, acmc-209y1n, 3-aminoazetidine dihy drochloric
FDPKMJDUXJFKOI-UHFFFAOYSA-N
azetidin-3-amine
1516506
95%
102065-89-4
MFCD09910173
Hygroscopic
Slightly soluble in water.
C1C(CN1)N
72.111
145.03

Identifiants chimiques

102065-89-4
72.111
FDPKMJDUXJFKOI-UHFFFAOYSA-N
1516506
C1C(CN1)N
C3H8N2
MFCD09910173
3-aminoazetidine, 3-azetidinamine, acmc-209y1n, 3-aminoazetidine dihy drochloric
azetidin-3-amine
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Offres spéciales

Offres spéciales

RUO – Research Use Only