Learn More
3-Aminobenzoic acid, 99+%, Thermo Scientific Chemicals
$54.14 - $265.83
Chemical Identifiers
CAS | 99-05-8 |
---|---|
Molecular Formula | C7H7NO2 |
Molecular Weight (g/mol) | 137.14 |
MDL Number | MFCD00007795 |
InChI Key | XFDUHJPVQKIXHO-UHFFFAOYSA-N |
Synonym | m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u |
PubChem CID | 7419 |
ChEBI | CHEBI:42682 |
IUPAC Name | 3-aminobenzoic acid |
SMILES | C1=CC(=CC(=C1)N)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC155850250
|
Thermo Scientific Chemicals
155850250 |
25 g |
Each for $54.14
|
|
|||||
AC155855000
|
Thermo Scientific Chemicals
155855000 |
500 g |
Each for $265.83
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-05-8 | |
137.14 | |
XFDUHJPVQKIXHO-UHFFFAOYSA-N | |
7419 | |
3-aminobenzoic acid |
C7H7NO2 | |
MFCD00007795 | |
m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u | |
CHEBI:42682 | |
C1=CC(=CC(=C1)N)C(=O)O |
Specifications
99-05-8 | |
100.0 | |
Beige-Brown to Brown-White | |
Authentic | |
Glass bottle | |
H2NC6H4CO2H | |
25 g | |
15, 416 | |
Solubility in water: 5.9g/L (15°C). Other solubilities: 3g/100g in ethanol (25°C),slightly soluble in ether | |
C1=CC(=CC(=C1)N)C(=O)O | |
137.14 | |
CHEBI:42682 | |
99+% | |
3-Aminobenzoic acid, 99+% |
99.0 | |
>172°C | |
150°C | |
99+% | |
C7H7NO2 | |
MFCD00007795 | |
14, 383 | |
m-aminobenzoic acid, m-carboxyaniline, benzoic acid, 3-amino, 3-carboxyaniline, maba, benzoic acid, m-amino, aniline-3-carboxylic acid, meta-aminobenzoic acid, m-aminobenzoesaeure, unii-g2x3b3o37u | |
XFDUHJPVQKIXHO-UHFFFAOYSA-N | |
3-aminobenzoic acid | |
7419 | |
137.14 | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 202-724-4
RTECSNumber : DG1225000
TSCA : TSCA
RUO – Research Use Only