missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzotrifluoride, 98%

CAS: 98-16-8 | C7H6F3N | 161.13 g/mol

$45.20 - $121.58

Chemical Identifiers

CAS 98-16-8
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.13
MDL Number MFCD00007797
InChI Key VIUDTWATMPPKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
PubChem CID 7375
IUPAC Name 3-(trifluoromethyl)aniline
SMILES C1=CC(=CC(=C1)N)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC103190050
SDS View Documents
Thermo Scientific Chemicals
103190050
5 mL Glass bottle
Each for $45.20
Only null left
 
AC103191000
SDS View Documents
Thermo Scientific Chemicals
103191000
100 mL Glass bottle
Each for $121.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-16-8
161.13
VIUDTWATMPPKEL-UHFFFAOYSA-N
7375
C1=CC(=CC(=C1)N)C(F)(F)F
C7H6F3N
MFCD00007797
3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
3-(trifluoromethyl)aniline

Specifications

98-16-8
5.0°C to 6.0°C
1.3030g/mL
85°C
97.5% min. (GC)
C7H6F3N
H2NC6H4CF3
5 mL
1.303
Solubility in water: 5g/L in water (20°C). Other solubilities: soluble in ethanol,ether and diethyl ether
C1=CC(=CC(=C1)N)C(F)(F)F
161.13
161.13
Liquid
98
Yellow
187.0°C to 193.0°C
Authentic
Glass bottle
1.4790 to 1.4810
MFCD00007797
12, 870
3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
VIUDTWATMPPKEL-UHFFFAOYSA-N
3-(trifluoromethyl)aniline
7375
98%
3-Aminobenzotrifluoride, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause damage to organs through prolo

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash w

GHS Signal Word: Danger

EINECSNumber : 202-643-4

RTECSNumber : XU9180000

TSCA : TSCA

Recommended Storage : This product may darken on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only