missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminocarbonylphenylboronic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 351422-73-6
Molecular Formula C7H8BNO3
Molecular Weight (g/mol) 164.96
MDL Number MFCD03411948
InChI Key WDGWHKRJEBENCE-UHFFFAOYSA-N
Synonym 3-aminocarbonylphenylboronic acid, 3-carbamoylphenyl boronic acid, benzamide-3-boronic acid, 3-carbamoylbenzeneboronic acid, 3-aminocarbonyphenylboronic acid, 3-aminocarbonyl benzeneboronic acid, 3-aminocarbonylphenyl boronic acid, 3-carboxamidephenylboronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonyl phenyl boronic acid
PubChem CID 2737814
IUPAC Name (3-carbamoylphenyl)boronic acid
SMILES NC(=O)C1=CC=CC(=C1)B(O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC444890010
SDS View Documents
Thermo Scientific Chemicals
444890010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

351422-73-6
164.96
WDGWHKRJEBENCE-UHFFFAOYSA-N
2737814
NC(=O)C1=CC=CC(=C1)B(O)O
C7H8BNO3
MFCD03411948
3-aminocarbonylphenylboronic acid, 3-carbamoylphenyl boronic acid, benzamide-3-boronic acid, 3-carbamoylbenzeneboronic acid, 3-aminocarbonyphenylboronic acid, 3-aminocarbonyl benzeneboronic acid, 3-aminocarbonylphenyl boronic acid, 3-carboxamidephenylboronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonyl phenyl boronic acid
(3-carbamoylphenyl)boronic acid

Specifications

351422-73-6
Glass bottle
MFCD03411948
3-aminocarbonylphenylboronic acid, 3-carbamoylphenyl boronic acid, benzamide-3-boronic acid, 3-carbamoylbenzeneboronic acid, 3-aminocarbonyphenylboronic acid, 3-aminocarbonyl benzeneboronic acid, 3-aminocarbonylphenyl boronic acid, 3-carboxamidephenylboronic acid, 3-aminocarbonyl phenylboronic acid, 3-aminocarbonyl phenyl boronic acid
NC(=O)C1=CC=CC(=C1)B(O)O
164.96
164.96
3-Aminocarbonylphenylboronic acid
96% min. (HPLC)
C7H8BNO3
1g
WDGWHKRJEBENCE-UHFFFAOYSA-N
(3-carbamoylphenyl)boronic acid
2737814
97%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only