missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopentane, 98%, Thermo Scientific Chemicals

$74.42 - $490.71

Chemical Identifiers

CAS 616-24-0
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00008096
InChI Key PQPFFKCJENSZKL-UHFFFAOYSA-N
Synonym 3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane
PubChem CID 12019
ChEBI CHEBI:84248
IUPAC Name pentan-3-amine
SMILES CCC(N)CC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0225006
View Documents
Thermo Scientific Chemicals
L0225006
5 g
Each for $74.42
 
AAL0225014
View Documents
Thermo Scientific Chemicals
L0225014
25 g
Each for $140.27
 
AAL0225022
View Documents
Thermo Scientific Chemicals
L0225022
100 g
Each for $490.71
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

616-24-0
87.17
PQPFFKCJENSZKL-UHFFFAOYSA-N
12019
pentan-3-amine
C5H13N
MFCD00008096
3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane
CHEBI:84248
CCC(N)CC

Specifications

616-24-0
89°C to 91°C
Amine-like
1.4055
5 g
635658
3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane
CCC(N)CC
87.17
CHEBI:84248
98%
0.748
2°C (35°F)
C5H13N
MFCD00008096
UN1106
Air Sensitive
PQPFFKCJENSZKL-UHFFFAOYSA-N
pentan-3-amine
12019
87.17
3-Aminopentane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES

EINECSNumber : 210-471-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only