Learn More
3-Aminopentane, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0225022
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Aminopentane | |
0.748 | |
2°C (35°F) | |
C5H13N | |
MFCD00008096 | |
UN1106 | |
Air Sensitive | |
PQPFFKCJENSZKL-UHFFFAOYSA-N | |
pentan-3-amine | |
12019 | |
87.17 |
616-24-0 | |
89°C to 91°C | |
Amine-like | |
1.4055 | |
100 g | |
635658 | |
3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane | |
CCC(N)CC | |
87.17 | |
CHEBI:84248 | |
98% |
Chemical Identifiers
616-24-0 | |
87.17 | |
PQPFFKCJENSZKL-UHFFFAOYSA-N | |
12019 | |
pentan-3-amine |
C5H13N | |
MFCD00008096 | |
3-aminopentane, 3-pentanamine, 1-ethylpropylamine, 3-pentylamine, propylamine, 1-ethyl, 3-amylamine, unii-3n2it605hv, 1-ethyl-propylamine, pent-3-ylamine, 3-amino pentane | |
CHEBI:84248 | |
CCC(N)CC |
Safety and Handling
GHS H Statement
H225-H314-H318-H302-H312-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYLAMINES
EINECSNumber : 210-471-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only