Learn More
3-Aminophthalhydrazide monosodium salt, 98+%, Thermo Scientific Chemicals
$96.74 - $354.48
Chemical Identifiers
CAS | 20666-12-0 |
---|---|
Molecular Formula | C8H9N3NaO2 |
Molecular Weight (g/mol) | 202.169 |
MDL Number | MFCD00161491 |
InChI Key | BTPIPBDXONOBET-UHFFFAOYSA-N |
Synonym | luminol sodium salt |
PubChem CID | 131673872 |
IUPAC Name | 5-amino-2,3-dihydrophthalazine-1,4-dione;molecular hydrogen;sodium |
SMILES | [HH].C1=CC2=C(C(=C1)N)C(=O)NNC2=O.[Na] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1520503
|
Thermo Scientific Chemicals
L1520503 |
1 g |
Each for $96.74
|
|
|||||
AAL1520506
|
Thermo Scientific Chemicals
L1520506 |
5 g |
Each for $354.48
|
|
|||||
Description
3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Aminophthalhydrazide monosodium salt is used for chemiluminescence analysis of metal cations, blood and glucococorticoids. It is also to image myeloperoxidase activity.
Solubility
Soluble in water (50 mg/ml).
Notes
Incompatible with bases, reducing agents and oxidizing agents. It is hygroscopic in nature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
20666-12-0 | |
202.169 | |
BTPIPBDXONOBET-UHFFFAOYSA-N | |
131673872 | |
[HH].C1=CC2=C(C(=C1)N)C(=O)NNC2=O.[Na] |
C8H9N3NaO2 | |
MFCD00161491 | |
luminol sodium salt | |
5-amino-2,3-dihydrophthalazine-1,4-dione;molecular hydrogen;sodium |
Specifications
20666-12-0 | |
MFCD00161491 | |
Hygroscopic | |
Soluble in water (50mg/ml). | |
[HH].C1=CC2=C(C(=C1)N)C(=O)NNC2=O.[Na] | |
202.169 | |
199.15 | |
3-Aminophthalhydrazide monosodium salt |
C8H9N3NaO2 | |
1 g | |
luminol sodium salt | |
BTPIPBDXONOBET-UHFFFAOYSA-N | |
5-amino-2,3-dihydrophthalazine-1,4-dione;molecular hydrogen;sodium | |
131673872 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 629-344-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only