Learn More
3-Aminopropionitrile, 98%, stab. with potassium carbonate, Thermo Scientific Chemicals
$75.08 - $690.30
Chemical Identifiers
CAS | 151-18-8 |
---|---|
Molecular Formula | C3H6N2 |
Molecular Weight (g/mol) | 70.10 |
MDL Number | MFCD00014820 |
InChI Key | AGSPXMVUFBBBMO-UHFFFAOYSA-N |
Synonym | 3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino |
PubChem CID | 1647 |
ChEBI | CHEBI:27413 |
IUPAC Name | 3-aminopropanenitrile |
SMILES | NCCC#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1304303
|
Thermo Scientific Chemicals
A1304303 |
1 g |
Each for $75.08
|
|
|||||
AAA1304306
|
Thermo Scientific Chemicals
A1304306 |
5 g |
Each for $201.08
|
|
|||||
AAA1304314
|
Thermo Scientific Chemicals
A1304314 |
25 g |
Each for $690.30
|
|
|||||
Description
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.
Solubility
Soluble in water.
Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
Chemical Identifiers
151-18-8 | |
70.10 | |
AGSPXMVUFBBBMO-UHFFFAOYSA-N | |
1647 | |
3-aminopropanenitrile |
C3H6N2 | |
MFCD00014820 | |
3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino | |
CHEBI:27413 | |
NCCC#N |
Specifications
151-18-8 | |
79°C to 81°C (16 mmHg) | |
C3H6N2 | |
H2NCH2CH2CN | |
1 g | |
1698848 | |
14,469 | |
Soluble in water. | |
NCCC#N | |
70.10 | |
CHEBI:27413 | |
98% |
0.952 | |
Amine-like | |
1.4375 | |
MFCD00014820 | |
UN2735 | |
Air and moisture sensitive | |
3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino | |
AGSPXMVUFBBBMO-UHFFFAOYSA-N | |
3-aminopropanenitrile | |
1647 | |
70.1 | |
3-Aminopropionitrile, stabilized with potassium carbonate |
Safety and Handling
GHS H Statement
H301-H314-H318-H361-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Combustible liquid.
P201-P202-P210-P235-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H314-H335-H361
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 205-786-0
RTECSNumber : UG0350000
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only